1-Chloro-3-fluorobenzene, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ100
162299-25G
162299-5G
SIAL162299-25GEA
1980
CZK
SIAL162299-25G
SIAL162299-5G
1-Chloro-3-fluorobenzene, Sigma-Aldrich®
1-Chloro-3-fluorobenzene
Vzorec:
C₆H₄ClF MW: 130,55 g/mol Bod varu: 126 °C (1013 hPa) Hustota: 1,221 g/cm³ (25 °C) |
MDL Number:
MFCD00000569 CAS číslo: 625-98-9 EINECS: 210-919-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...