Tisk…

4-Amino-5-chloro-2-methoxybenzoic acid, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: Metoclopramide Impurity C, 4-Amino-5-chloro-o-anisic acid

M-Clarity™ quality level = MQ100

340871-25G 340871-5G
SIAL340871-25GEA 2900 CZK
SIAL340871-25G SIAL340871-5G
4-Amino-5-chloro-2-methoxybenzoic acid, Sigma-Aldrich®
4-Amino-5-chloro-2-methoxybenzoic acid
Vzorec: C₈H₈ClNO₃
Bod varu: 354 °C (1013 hPa)
Bod tání: 213…215 °C
MDL Number: MFCD00038794
CAS číslo: 7206-70-4
EINECS: 230-582-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR