Methyl-4-chlorobenzoate, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
4-Chlorobenzoic acid emthyl ester, Methyl 4-chlorobenzoate
M-Clarity™ quality level = MQ100
216089-5G
216089-100G
SIAL216089-5GEA
1430
CZK
SIAL216089-5G
SIAL216089-100G
Methyl-4-chlorobenzoate, Sigma-Aldrich®
Methyl-4-chlorobenzoate
Vzorec:
C₈H₇ClO₂ MW: 170,6 g/mol Bod varu: 103…106 °C (15 mmHg) Bod tání: 43 °C Hustota: 1,382 g/cm³ (20 °C) |
MDL Number:
MFCD00000621 CAS číslo: 1126-46-1 EINECS: 214-420-9 Merck Index: 13,02144 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...