Tisk…

Methyl-3-chlorobenzoate, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: 3-Chlorobenzoic acid emthyl ester, Methyl 3-chlorobenzoate

M-Clarity™ quality level = MQ100

234621-25G 234621-5G
SIAL234621-25GEA 3800 CZK
SIAL234621-25G SIAL234621-5G
Methyl-3-chlorobenzoate, Sigma-Aldrich®
Methyl-3-chlorobenzoate
Vzorec: C₈H₇ClO₂
MW: 170,6 g/mol
Bod varu: 231 °C (1013 hPa)
Bod tání: 21 °C
Hustota: 1,23 g/cm³ (20 °C)
MDL Number: MFCD00000592
CAS číslo: 2905-65-9
EINECS: 220-814-1
Merck Index: 13,02142

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR