Tisk…

5-Chloro-2-nitrobenzamide ≥98%

Dodavatel: Thermo Fisher Scientific
Varování

L09380.09 L09380.04
L09380.09EA 2120 CZK
L09380.09 L09380.04
5-Chloro-2-nitrobenzamide ≥98%
5-Chloro-2-nitrobenzamide

Purity: 98+% CAS No.: 40763-96-0 Molecular Formula: C7H5ClN2O3 Formula Weight: 200.58 Melting Point: 155-159° Beilstein Reference: 2267797

Vzorec: C₇H₅ClN₂O₃
Bod tání: 155…159 °C
Storage Temperature: Ambient
MDL Number: MFCD00017509
CAS číslo: 40763-96-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR