Tisk…

2-Chloro-4-nitrobenzamide ≥98%

Dodavatel: Thermo Scientific
Nebezpečí

Synonyms: Aklomide

A19702.06 A19702.14
A19702.06EA 1850 CZK
A19702.06 A19702.14
2-Chloro-4-nitrobenzamide ≥98%
2-Chloro-4-nitrobenzamide

Purity: 98% CAS No.: 3011-89-0 Molecular Formula: C7H5ClN2O3 Formula Weight: 200.58 Melting Point: 168-172° Beilstein Reference: 1966537

Vzorec: C₇H₅ClN₂O₃
Bod tání: 168…172 °C
Storage Temperature: Ambient
MDL Number: MFCD00017119
CAS číslo: 3011-89-0
EINECS: 221-143-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR