Tisk…

2,6-Dichlorobenzamide ≥98%

Dodavatel: Thermo Scientific

A10865.06 A10865.14 A10865.22
A10865.06EA 824 CZK
A10865.06 A10865.14 A10865.22
2,6-Dichlorobenzamide ≥98%
2,6-Dichlorobenzamide

Purity: 98% CAS No.: 2008-58-4 Molecular Formula: C7H5Cl2NO Formula Weight: 190.03 Melting Point: 198-200° Beilstein Reference: 1869103

Vzorec: C₇H₅Cl₂NO
Bod varu: 335 °C (1013 hPa)
Bod tání: 198…200 °C
Hustota: 1,44 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00007975
CAS číslo: 2008-58-4
EINECS: 217-918-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR