Tisk…

4-Chlorobenzoyl chloride ≥98%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

Synonyms: p-Chlorobenzoyl chloride

A16325.14 A16325.22 A16325.36
A16325.14EA 783 CZK
A16325.14 A16325.22 A16325.36
4-Chlorobenzoyl chloride ≥98%
4-Chlorobenzoyl chloride

Purity: 98% CAS No.: 122-01-0 Molecular Formula: C7H4Cl2O Formula Weight: 175.01 Flash Point: 105°(221°F) Refractive Index: 1.5780 Odour: Acrid Melting Point: 12-14° Boiling Point: 221-222° Beilstein Reference: 471606 UN No.: 3265 MDL No.: MFCD00000686

Vzorec: C₇H₄Cl₂O
Bod varu: 221…222 °C
Bod tání: 12…14 °C
Hustota: 1,377
Bod vzplanutí: 105 °C (221 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000686
CAS číslo: 122-01-0
EINECS: 204-515-3
UN: 3265
ADR: 8,II

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR