3-Chlorobenzoyl chloride ≥97%
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
Synonyms:
m-Chlorobenzoyl chloride, 3-Chlorobenzoylchloride
A13509.14
A13509.22
A13509.14EA
1300
CZK
A13509.14
A13509.22
3-Chlorobenzoyl chloride ≥97%
3-Chlorobenzoyl chloride
Purity: 97%
CAS No.: 618-46-2
Molecular Formula: C7H4Cl2O
Formula Weight: 175.01
Flash Point: 117°(242°F)
Refractive Index: 1.5690
Odour: Acidic
Boiling Point: 224-225°
Beilstein Reference: 386256
UN No.: 3265
MDL No.: MFCD00000671
Vzorec:
C₇H₄Cl₂O Bod varu: 224…225 °C Bod tání: 44 °C Hustota: 1,368 Bod vzplanutí: 117 °C (242 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000671 CAS číslo: 618-46-2 EINECS: 210-552-6 UN: 3265 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...