Tisk…

3,5-Dichlorobenzoyl chloride 96%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

B24676.06 B24676.14 B24676.22
B24676.06EA 1010 CZK
B24676.06 B24676.14 B24676.22
3,5-Dichlorobenzoyl chloride 96%
3,5-Dichlorobenzoyl chloride

Purity: 96% CAS No.: 2905-62-6 Molecular Formula: C7H3Cl3O Formula Weight: 209.46 Flash Point: 137°(278°F) Refractive Index: 1.5820 Melting Point: 24-28° Boiling Point: 245° Beilstein Reference: 1940681 UN No.: 3261 MDL No.: MFCD00009817

Vzorec: C₇H₃Cl₃O
Bod varu: 245 °C
Bod tání: 24…28 °C
Hustota: 1,478
Bod vzplanutí: 137 °C (278 °F)
Storage Temperature: Ambient
MDL Number: MFCD00009817
CAS číslo: 2905-62-6
EINECS: 220-813-6
UN: 3261
ADR: 8,II

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR