2,4,6-Trichlorobenzoyl chloride ≥98%
Dodavatel: Thermo Scientific
|
Nebezpečí
|
Synonyms:
2,4,6-Trichlorobenzoyl Chloride
L14159.06
L14159.14
L14159.06EA
1800
CZK
L14159.06
L14159.14
2,4,6-Trichlorobenzoyl chloride ≥98%
2,4,6-Trichlorobenzoyl chloride
Purity: 98%
CAS No.: 4136-95-2
Molecular Formula: C7H2Cl4O
Formula Weight: 243.91
Flash Point:
Vzorec:
C₇H₂Cl₄O MW: 243,9 g/mol Bod varu: 107…108 °C (6 mmHg) Hustota: 1,561 Bod vzplanutí: >110 °C (230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00075323 CAS číslo: 4136-95-2 UN: 3265 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...