Tisk…

2,3-Dichlorobenzoyl chloride ≥98%

Dodavatel: Thermo Scientific
Nebezpečí

L09506.06 L09506.14 L09506.22
L09506.06EA 1460 CZK
L09506.06 L09506.14 L09506.22
2,3-Dichlorobenzoyl chloride ≥98%
2,3-Dichlorobenzoyl chloride

Purity: 98% CAS No.: 2905-60-4 Molecular Formula: C7H3Cl3O Formula Weight: 209.46 Flash Point: 167°(332°F) Odour: Pungent Boiling Point: 140°/14mm Beilstein Reference: 2575973 UN No.: 3261 MDL No.: MFCD00035937

Vzorec: C₇H₃Cl₃O
MW: 209,46 g/mol
Bod varu: 140 °C (14 mmHg)
Bod tání: 28…32 °C
Bod vzplanutí: 167 °C (332 °F)
Storage Temperature: Ambient
MDL Number: MFCD00035937
CAS číslo: 2905-60-4
EINECS: 220-811-5
UN: 3261
ADR: 8,II

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR