5-Chloro-2-nitrobenzaldehyde, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ100
C58800-25G
C58800-5G
SIALC58800-25GEA
11930
CZK
SIALC58800-25G
SIALC58800-5G
5-Chloro-2-nitrobenzaldehyde, Sigma-Aldrich®
5-Chloro-2-nitrobenzaldehyde
Vzorec:
C₇H₄ClNO₃ MW: 185,57 g/mol Bod tání: 77,5 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007289 CAS číslo: 6628-86-0 EINECS: 229-614-9 UN: 3077 ADR: 9,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...