2,5-Dichlorophenylboronic acid ≥98%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
B22984.03
B22984.06
B22984.14
B22984.03EA
1160
CZK
B22984.03
B22984.06
B22984.14
2,5-Dichlorophenylboronic acid ≥98%
2,5-Dichlorophenylboronic acid
Purity: 98%
CAS No.: 135145-90-3
Molecular Formula: C6H5BCl2O2
Formula Weight: 190.82
Melting Point: ca 210-216°
Beilstein Reference: 4801290
Vzorec:
C₆H₅BCl₂O₂ Bod tání: ∼ 210…216 °C Storage Temperature: Ambient |
MDL Number:
MFCD01863182 CAS číslo: 135145-90-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...