2,3-Dichlorophenylboronic acid, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ200
514047-25G
514047-5G
SIAL514047-25GEA
0
CZK
SIAL514047-25G
SIAL514047-5G
2,3-Dichlorophenylboronic acid, Sigma-Aldrich®
2,3-Dichlorophenylboronic acid
Vzorec:
C₆H₅BCl₂O₂ MW: 190,82 g/mol |
MDL Number:
MFCD01075703 CAS číslo: 151169-74-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...