Tisk…

Terephthalaldehyde ≥98%

Dodavatel: Thermo Scientific
Varování

Synonyms: Terephthaldialdehyde, Terephthaldicarboxaldehyde, p-Phthalaldehyde

A14930.14 A14930.22 A14930.36
A14930.14EA 1620 CZK
A14930.14 A14930.22 A14930.36
Terephthalaldehyde ≥98%
Terephthalaldehyde

Purity: 98+% CAS No.: 623-27-8 Molecular Formula: C8H6O2 Formula Weight: 134.14 Flash Point: 76°(168°F) Melting Point: 114-117° Boiling Point: 245° Beilstein Reference: 385863 MDL No.: MFCD00006949

Vzorec: C₈H₆O₂
MW: 134,13 g/mol
Bod varu: 245 °C
Bod tání: 114…117 °C
Hustota: 1,060
Bod vzplanutí: 76 °C (168 °F)
Storage Temperature: Ambient
MDL Number: MFCD00006949
CAS číslo: 623-27-8
EINECS: 210-784-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR