Tisk…

Syringaldehyde, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: 3,5-Dimethoxy-4-hydroxybenzaldehyde, 4-Hydroxy-3,5-dimethoxybenzaldehyde

M-Clarity™ quality level = MQ300

W404926-100G W404926-1KG W404926-5KG W404926-SAMPLE
SIALW404926-100GEA 4820 CZK
SIALW404926-100G SIALW404926-1KG SIALW404926-5KG SIALW404926-SAMPLE
Syringaldehyde, Sigma-Aldrich®
Syringaldehyde
Vzorec: C₉H₁₀O₄
MW: 182,18 g/mol
Bod varu: 322 °C (1013 hPa)
Bod tání: 107 °C
Storage Temperature: Ambient
MDL Number: MFCD00006943
CAS číslo: 134-96-3
EINECS: 205-167-5
Merck Index: 12,09191

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR