3,5-Dimethoxybenzaldehyde, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ100
126292-10G
126292-50G
SIAL126292-10GEA
4420
CZK
SIAL126292-10G
SIAL126292-50G
3,5-Dimethoxybenzaldehyde, Sigma-Aldrich®
3,5-Dimethoxybenzaldehyde
Vzorec:
C₉H₁₀O₃ MW: 166,18 g/mol Bod varu: 277 °C (1013 hPa) Bod tání: 48 °C Storage Temperature: Ambient |
MDL Number:
MFCD00003366 CAS číslo: 7311-34-4 EINECS: 230-772-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...