2,5-Dimethoxybenzaldehyde, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ100
D130605-100G
D130605-25G
SIALD130605-100GEA
2170
CZK
SIALD130605-100G
SIALD130605-25G
2,5-Dimethoxybenzaldehyde, Sigma-Aldrich®
2,5-Dimethoxybenzaldehyde
Vzorec:
C₉H₁₀O₃ MW: 166,18 g/mol Bod varu: 284 °C (1013 hPa) Bod tání: 49…51 °C Storage Temperature: Ambient |
MDL Number:
MFCD00003314 CAS číslo: 93-02-7 EINECS: 202-211-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...