Tisk…

5-Methoxysalicylaldehyde, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: 6-Hydroxy-m-anisaldehyde, 2-Hydroxy-5-methoxybenzaldehyde

M-Clarity™ quality level = MQ100

146862-1G 146862-5G
SIAL146862-1GEA 1010 CZK
SIAL146862-1G SIAL146862-5G
5-Methoxysalicylaldehyde, Sigma-Aldrich®
5-Methoxysalicylaldehyde
Vzorec: C₈H₈O₃
MW: 152.15
Bod varu: 248
Bod tání: 51…52 °C
Hustota: 1.21 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003332
CAS číslo: 672-13-9
EINECS: 211-589-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR