o-Anisaldehyde, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ100
109622-100G
109622-2G
109622-500G
SIAL109622-100GEA
1840
CZK
SIAL109622-100G
SIAL109622-2G
SIAL109622-500G
o-Anisaldehyde, Sigma-Aldrich®
o-Anisaldehyde
Vzorec:
C₈H₈O₂ Bod varu: 238 °C (1013 hPa) Bod tání: 34…37 °C Hustota: 1,127 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00003308 CAS číslo: 135-02-4 EINECS: 205-171-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...