Tisk…

5-Hydroxy-2-nitrobenzaldehyde, Sigma-Aldrich®

Dodavatel: Merck
Varování


M-Clarity™ quality level = MQ200

H48107-25G H48107-5G
SIALH48107-25GEA 10570 CZK
SIALH48107-25G SIALH48107-5G
5-Hydroxy-2-nitrobenzaldehyde, Sigma-Aldrich®
5-Hydroxy-2-nitrobenzaldehyde
Vzorec: C₇H₅NO₄
MW: 167,12 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00007332
CAS číslo: 42454-06-8
EINECS: 255-832-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR