Tisk…

3-Methoxy-2-nitrobenzaldehyde, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

163821-25G 163821-5G
SIAL163821-25GEA 37860 CZK
SIAL163821-25G SIAL163821-5G
3-Methoxy-2-nitrobenzaldehyde, Sigma-Aldrich®
3-Methoxy-2-nitrobenzaldehyde
Vzorec: C₈H₇NO₄
MW: 181,15 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00007135
CAS číslo: 53055-05-3
EINECS: 258-332-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR