4-Cyanobenzaldehyde, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
4-Formylbenzonitrile
M-Clarity™ quality level = MQ100
C89609-1G
C89609-25G
C89609-5G
SIALC89609-1GEA
945
CZK
SIALC89609-1G
SIALC89609-25G
SIALC89609-5G
4-Cyanobenzaldehyde, Sigma-Aldrich®
4-Cyanobenzaldehyde
Vzorec:
C₈H₅NO Bod varu: 262 °C (1013 hPa) Bod tání: 100…103 °C Storage Temperature: Ambient |
MDL Number:
MFCD00003376 CAS číslo: 105-07-7 EINECS: 203-267-3 UN: 3439 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...