3-Bromobenzenesulphonyl chloride, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ100
545716-1G
545716-5G
SIAL545716-1GEA
2260
CZK
SIAL545716-1G
SIAL545716-5G
3-Bromobenzenesulphonyl chloride, Sigma-Aldrich®
3-Bromobenzenesulphonyl chloride
Vzorec:
C₆H₄BrClO₂S MW: 255,52 g/mol Bod varu: 95…95,5 °C (0.8 mmHg) Bod tání: 31…33 °C Hustota: 1,773 g/cm³ (20 °C) |
MDL Number:
MFCD00052313 CAS číslo: 2905-24-0 UN: 3261 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...