3-Bromobenzenesulphonyl chloride ≥97%
Dodavatel: Thermo Scientific
|
Nebezpečí
|
A11507.03
A11507.06
A11507.14
A11507.03EA
1680
CZK
A11507.03
A11507.06
A11507.14
3-Bromobenzenesulphonyl chloride ≥97%
3-Bromobenzenesulphonyl chloride
Purity: 97%
CAS No.: 2905-24-0
Molecular Formula: C6H4BrClO2S
Formula Weight: 255.52
Flash Point:
Vzorec:
C₆H₄BrClO₂S MW: 255,52 g/mol Bod varu: 90…91 °C (0,5 mmHg) Bod tání: 30…33 °C Hustota: 1,773 Bod vzplanutí: >110 °C (230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00052313 CAS číslo: 2905-24-0 UN: 3261 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...