2-Bromobenzenesulphonyl chloride, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
2-Bromobenzene-1-sulphonyl chloride
M-Clarity™ quality level = MQ100
442844-1G
442844-5G
SIAL442844-1GEA
1130
CZK
SIAL442844-1G
SIAL442844-5G
2-Bromobenzenesulphonyl chloride, Sigma-Aldrich®
2-Bromobenzenesulphonyl chloride
Vzorec:
C₆H₄BrClO₂S MW: 255,52 g/mol Bod varu: 127…128 °C (3,6 mmHg) Bod tání: 51 °C |
MDL Number:
MFCD00051971 CAS číslo: 2905-25-1 UN: 3261 ADR: 8,II |
Learn more
![](/m-cz.vwr.com/cs_CZ/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...