Tisk…

2-Bromothioanisole ≥98%

Dodavatel: Thermo Scientific
Varování

Synonyms: 1-Bromo-2-(methylthio)benzene, 2-Bromophenyl methyl sulphide, (2-Bromophenyl)(methyl)sulphane

A13632.03 A13632.06 A13632.14
A13632.03EA 1020 CZK
A13632.03 A13632.06 A13632.14
2-Bromothioanisole ≥98%
2-Bromothioanisole

Purity: 98% CAS No.: 19614-16-5 Molecular Formula: C7H7BrS Formula Weight: 203.11 Flash Point: 110°(230°F) Refractive Index: 1.6330 Melting Point: -24° Boiling Point: 140-141°/20mm Beilstein Reference: 2243716 MDL No.: MFCD00000066

Vzorec: C₇H₇BrS
MW: 203,1 g/mol
Bod varu: 140…141 °C (20 mmHg)
Bod tání: –24 °C
Hustota: 1,524
Bod vzplanutí: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000066
CAS číslo: 19614-16-5
EINECS: 243-183-4
UN: 3334
ADR: 9,

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR