4-Bromoveratrole, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
1-Bromo-3,4-dimethoxybenzene, 4-Bromo-1,2-dimethoxybenzene
M-Clarity™ quality level = MQ100
B83355-100G
B83355-25G
SIALB83355-100GEA
5550
CZK
SIALB83355-100G
SIALB83355-25G
4-Bromoveratrole, Sigma-Aldrich®
4-Bromoveratrole
Vzorec:
C₈H₉BrO₂ MW: 217.06 Bod varu: 255 Hustota: 1.5068 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00008381 CAS číslo: 2859-78-1 EINECS: 220-677-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...