3-Bromo-N,N-dimethylaniline, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ100
666939-1G
666939-5G
SIAL666939-1GEA
1250
CZK
SIAL666939-1G
SIAL666939-5G
3-Bromo-N,N-dimethylaniline, Sigma-Aldrich®
3-Bromo-N,N-dimethylaniline
Vzorec:
C₈H₁₀BrN MW: 200,08 g/mol Bod varu: 70 °C (0,4 mmHg) Bod tání: 11 °C Hustota: 1,3652 g/cm³ (19 °C) |
MDL Number:
MFCD00045020 CAS číslo: 16518-62-0 |
Learn more
![](/m-cz.vwr.com/cs_CZ/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...