3-Bromo-5-iodobenzoic acid ≥97%
Dodavatel: Thermo Scientific
|
Varování
|
A17076.14
A17076.06
A17076.22
A17076.14EA
6560
CZK
A17076.14
A17076.06
A17076.22
3-Bromo-5-iodobenzoic acid ≥97%
3-Bromo-5-iodobenzoic acid
Purity: 97%
CAS No.: 188815-32-9
Molecular Formula: C7H4BrIO2
Formula Weight: 326.92
Melting Point: 218-221°
Beilstein Reference: 3240837
Vzorec:
C₇H₄BrIO₂ MW: 326,92 g/mol Bod tání: 218…221 °C Storage Temperature: Ambient |
MDL Number:
MFCD00191851 CAS číslo: 188815-32-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...