2-Bromo-4-fluoroaniline ≥98%
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
A11751.06
A11751.14
A11751.06EA
1360
CZK
A11751.06
A11751.14
2-Bromo-4-fluoroaniline ≥98%
2-Bromo-4-fluoroaniline
Purity: 98%
CAS No.: 1003-98-1
Molecular Formula: C6H5BrFN
Formula Weight: 190.01
Flash Point: 104°(219°F)
Refractive Index: 1.5830
Boiling Point: 221°
Beilstein Reference: 2802562
UN No.: 2810
MDL No.: MFCD00042462
Vzorec:
C₆H₅BrFN Bod varu: 221 °C Bod tání: 41 °C Hustota: 1,670 Bod vzplanutí: 104 °C (219 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00042462 CAS číslo: 1003-98-1 UN: 2810 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...