Tisk…

2-Brom-4,6-difluoranilin ≥98%

Dodavatel: Thermo Fisher Scientific
Varování

A11022.14 A11022.22 A11022.36
A11022.14EA 1500 CZK
A11022.14 A11022.22 A11022.36
2-Brom-4,6-difluoranilin ≥98%
2-Brom-4,6-difluoranilin

Purity: 98% CAS No.: 444-14-4 Molecular Formula: C6H4BrF2N Formula Weight: 208.01 Odour: Weak, characteristic Melting Point: 38-42° Beilstein Reference: 2718139 UN No.: 3077 MDL No.: MFCD00009639

Vzorec: C₆H₄BrF₂N
Bod varu: 227 °C (1013 hPa)
Bod tání: 38…42 °C
Storage Temperature: Ambient
MDL Number: MFCD00009639
CAS číslo: 444-14-4
EINECS: 429-430-0
UN: 3077
ADR: 9,III

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR