Bromopentafluorobenzene, Sigma-Aldrich®
Dodavatel: Merck
|
Varování
|
Synonyms:
1-Bromo-2,3,4,5,6-pentafluorobenzene
M-Clarity™ quality level = MQ100
B75158-10G
B75158-50G
SIALB75158-10GEA
2250
CZK
SIALB75158-10G
SIALB75158-50G
Bromopentafluorobenzene, Sigma-Aldrich®
Bromopentafluorobenzene
Vzorec:
C₆BrF₅ MW: 246.96 Bod varu: 137 Bod tání: –31 °C Hustota: 1.9503 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00000287 CAS číslo: 344-04-7 EINECS: 206-449-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...