Tisk…

Calcium L-lactate hydrate ~90%, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: Sarcolactic acid hemicalcium salt, L-Lactic acid hemicalcium salt, (S)-(+)-2-Hydroxypropanoic acid hemicalcium salt

L2000-1KG L2000-250G
SIALL2000-1KGEA 2910 CZK
SIALL2000-1KG SIALL2000-250G
Calcium L-lactate hydrate ~90%, Sigma-Aldrich®
Calcium L-lactate hydrate

Calcium L-lactate hydrate has been used as a sole carbon and energy source, to study the sulfate reduction ability of Desulfovibrio desulfuricans under different conditions.

Vzorec: C₆H₁₀CaO₆.xH₂O
Storage Temperature: Refrigerator
MDL Number: MFCD00150344
CAS číslo: 41372-22-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR