Trimethylolpropane triglycidyl ether, technická kvalita, Sigma-Aldrich®
Dodavatel: Merck
|
Nebezpečí
|
Synonyms:
1,1,1-Tris(glycidyloxymethyl)propane, 2-({2,2-BIS[(OXIRAN-2-YLMETHOXY)METHYL]BUTOXY}METHYL)OXIRANE, 1-(2,3-Epoxypropoxy)-2,2-bis((2,3-epoxypropoxy)methyl)butane
M-Clarity™ quality level = MQ200
430269-1L
430269-250ML
SIAL430269-1LEA
9500
CZK
SIAL430269-1L
SIAL430269-250ML
Trimethylolpropane triglycidyl ether, technická kvalita, Sigma-Aldrich®
Trimethylolpropane triglycidyl ether
Additive in fast-setting adhesives, low temperature curing coatings and hard, abrasion resistant castings and decoupage systems.
Reduces viscosity but retains high epoxide level, imparts hardness and toughness to epoxide polymers. Improves solubility/compatibility characteristics of highly aromatic epoxy systems.
Vzorec:
C₁₅H₂₆O₆ MW: 302,36 g/mol Hustota: 1,157 g/ml (25 °C) (lit.) Bod vzplanutí: 113 °C |
MDL Number:
MFCD00274208 CAS číslo: 30499-70-8 |
Výsledky specifikačního testu
Infrared Spectrum | Conforms to Structure |
Viscosity (Ubbelohde at 25 °C) | 120 - 180 cps |
Colour Test (Colour Gardner) | ≤2 |
Miscellaneous Assay, Weight per Epoxide (g/Eq) | 138 - 154 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...