1-Bromo-2-fluorobenzene, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ200
B66809-100G
B66809-25G
SIALB66809-100GEA
4120
CZK
SIALB66809-100G
SIALB66809-25G
1-Bromo-2-fluorobenzene, Sigma-Aldrich®
1-Bromo-2-fluorobenzene
Vzorec:
C₆H₄BrF Bod varu: 155 °C (1013 hPa) Hustota: 1,61 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00000282 CAS číslo: 1072-85-1 EINECS: 214-018-3 UN: 1993 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...