4-Bromo-3,5-dimethoxybenzoic acid, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ100
579718-25G
579718-5G
SIAL579718-25GEA
4880
CZK
SIAL579718-25G
SIAL579718-5G
4-Bromo-3,5-dimethoxybenzoic acid, Sigma-Aldrich®
4-Bromo-3,5-dimethoxybenzoic acid
Vzorec:
C₉H₉BrO₄ MW: 261,07 g/mol |
MDL Number:
MFCD01632140 CAS číslo: 56518-42-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...