Tisk…

2-Bromo-4,5-dimethoxybenzoic acid, Sigma-Aldrich®

Dodavatel: Merck
Varování

Synonyms: 6-Bromoveratric acid

M-Clarity™ quality level = MQ100

441074-1G 441074-5G
SIAL441074-1GEA 620 CZK
SIAL441074-1G SIAL441074-5G
2-Bromo-4,5-dimethoxybenzoic acid, Sigma-Aldrich®
2-Bromo-4,5-dimethoxybenzoic acid
Vzorec: C₉H₉BrO₄
MW: 261,07 g/mol
MDL Number: MFCD00017542
CAS číslo: 6286-46-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR