Tisk…

5-Bromo-2-methoxybenzoic acid, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

694789-1G 694789-5G
SIAL694789-1GEA 1590 CZK
SIAL694789-1G SIAL694789-5G
5-Bromo-2-methoxybenzoic acid, Sigma-Aldrich®
5-Bromo-2-methoxybenzoic acid
Vzorec: C₈H₇BrO₃
MW: 231,05 g/mol
MDL Number: MFCD00093713
CAS číslo: 2476-35-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR