Ethyl-4-bromobenzoate, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
4-Bromobenzoic acid ethyl ester, Ethyl 4-bromobenzoate
M-Clarity™ quality level = MQ200
363774-100G
363774-25G
SIAL363774-100GEA
7950
CZK
SIAL363774-100G
SIAL363774-25G
Ethyl-4-bromobenzoate, Sigma-Aldrich®
Ethyl-4-bromobenzoate
Vzorec:
C₉H₉BrO₂ Bod varu: 264 °C (1013 hPa) Bod tání: 18 °C Hustota: 1,4332 g/cm³ (17.3 °C) |
MDL Number:
MFCD00016329 CAS číslo: 5798-75-4 EINECS: 227-347-2 Merck Index: 13,01392 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...