Tisk…

Methyl-2-bromobenzoate, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: 2-Bromobenzoic acid methyl ester, Methyl 2-bromobenzoate

M-Clarity™ quality level = MQ200

387851-25ML 387851-5ML
SIAL387851-25MLEA 1650 CZK
SIAL387851-25ML SIAL387851-5ML
Methyl-2-bromobenzoate, Sigma-Aldrich®
Methyl-2-bromobenzoate
Vzorec: C₈H₇BrO₂
MW: 215,05 g/mol
Bod varu: 253 °C (1013 hPa)
Hustota: 1,51 g/cm³ (20 °C)
MDL Number: MFCD00016328
CAS číslo: 610-94-6
EINECS: 210-241-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR