4-Bromobenzoyl chloride, Sigma-Aldrich®
Dodavatel: Merck
|
Nebezpečí
|
Synonyms:
p-Bromobenzoyl chloride
M-Clarity™ quality level = MQ100
B59209-10G
B59209-25G
SIALB59209-10GEA
1640
CZK
SIALB59209-10G
SIALB59209-25G
4-Bromobenzoyl chloride, Sigma-Aldrich®
4-Bromobenzoyl chloride
Vzorec:
C₇H₄BrClO Bod varu: 247 °C (1013 hPa) Bod tání: 36 °C Bod vzplanutí: >110 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00000683 CAS číslo: 586-75-4 EINECS: 209-580-1 UN: 3261 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...