2-Bromo-5-chloroanisole ≥98%
Dodavatel: Thermo Scientific
Synonyms:
1-Bromo-4-chloro-2-methoxybenzene
B25512.06
B25512.14
B25512.06EA
3210
CZK
B25512.06
B25512.14
2-Bromo-5-chloroanisole ≥98%
2-Bromo-5-chloroanisole
Purity: 98+%
CAS No.: 174913-09-8
Molecular Formula: C7H6BrClO
Formula Weight: 221.48
Refractive Index: 1.5875
Vzorec:
C₇H₆BrClO MW: 221,48 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD03790889 CAS číslo: 174913-09-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...