1-Bromo-3-chloro-5-fluorobenzene ≥98%
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
3-Chloro-5-fluorobromobenzene
A16597.06
A16597.14
A16597.06EA
1190
CZK
A16597.06
A16597.14
1-Bromo-3-chloro-5-fluorobenzene ≥98%
1-Bromo-3-chloro-5-fluorobenzene
Purity: 98%
CAS No.: 33863-76-2
Molecular Formula: C6H3BrClF
Formula Weight: 209.45
Refractive Index: 1.5470
Beilstein Reference: 2432912
Vzorec:
C₆H₃BrClF MW: 209,44 g/mol Hustota: 1,72 Storage Temperature: Ambient |
MDL Number:
MFCD00070747 CAS číslo: 33863-76-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...