3-Bromophenylboronic acid ≥98%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
L16354.06
L16354.14
L16354.06EA
2050
CZK
L16354.06
L16354.14
3-Bromophenylboronic acid ≥98%
3-Bromophenylboronic acid
Purity: 98+%
CAS No.: 89598-96-9
Molecular Formula: C6H6BBrO2
Formula Weight: 200.83
Melting Point: 164-168°
Beilstein Reference: 3127104
Vzorec:
C₆H₆BBrO₂ Bod tání: 164…168 °C Storage Temperature: Ambient |
MDL Number:
MFCD00239386 CAS číslo: 89598-96-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...