Tisk…

3,5-Dibromophenylboronic acid ≥97%

Dodavatel: Thermo Scientific
Varování

B23863.06 B23863.14
B23863.06EA 3810 CZK
B23863.06 B23863.14
3,5-Dibromophenylboronic acid ≥97%
3,5-Dibromophenylboronic acid

Purity: 97% CAS No.: 117695-55-3 Molecular Formula: C6H5BBr2O2 Formula Weight: 279.73 Melting Point: 300° Beilstein Reference: 6645285

Vzorec: C₆H₅BBr₂O₂
MW: 279,72 g/mol
Bod tání: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD01075725
CAS číslo: 117695-55-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR