Tisk…

4-Hydroxyphenylboronic acid, Sigma-Aldrich®

Dodavatel: Merck
Varování

Synonyms: (4-Hydroxyphenyl)metaboric acid, Dihydroxy(4-hydroxyphenyl)borane, 4-Hydroxybenzeneboronic acid

M-Clarity™ quality level = MQ100

523976-1G 523976-5G
SIAL523976-1GEA 1630 CZK
SIAL523976-1G SIAL523976-5G
4-Hydroxyphenylboronic acid, Sigma-Aldrich®
4-Hydroxyphenylboronic acid
Vzorec: C₆H₇BO₃
MW: 137,93 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01074628
CAS číslo: 71597-85-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR