Tisk…

3-Hydroxyphenylboronic acid, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: (3-Hydroxyphenyl)metaboric acid, (3-Hydroxyphenyl)boronic acid, Dihydroxy(3-hydroxyphenyl)borane, 3-Hydroxybenzeneboronic acid

M-Clarity™ quality level = MQ100

523968-10G 523968-1G
SIAL523968-10GEA 8960 CZK
SIAL523968-10G SIAL523968-1G
3-Hydroxyphenylboronic acid, Sigma-Aldrich®
3-Hydroxyphenylboronic acid
Vzorec: C₆H₇BO₃
MW: 137,93 g/mol
MDL Number: MFCD01074603
CAS číslo: 87199-18-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR