3,4,5-Trimethoxyphenylboronic acid ≥98%
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
3,4,5-Trimethoxybenzeneboronic acid, Dihydroxy(3,4,5-trimethoxyphenyl)borane, (3,4,5-Trimethoxyphenyl)metaboric acid
L15191.03
L15191.06
L15191.03EA
1980
CZK
L15191.03
L15191.06
3,4,5-Trimethoxyphenylboronic acid ≥98%
3,4,5-Trimethoxyphenylboronic acid
Purity: 98%
CAS No.: 182163-96-8
Molecular Formula: C9H13BO5
Formula Weight: 212.01
Melting Point: ca 245°
Beilstein Reference: 8205051
Vzorec:
C₉H₁₃BO₅ MW: 212,01 g/mol Bod tání: ∼ 245 °C Storage Temperature: Ambient |
MDL Number:
MFCD01075676 CAS číslo: 182163-96-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...